Key Documents
502693
Upchurch Precolumn MicroFilter
for use with 1/16 in. O.D. tubing, PEEK frit
About This Item
UNSPSC Code:
41115700
material
PEEK frit
packaging
pkg of 1 ea
fitting
10-32 female Upchurch inlet fitting for 1/16 in. O.D. tubing
10-32 male Upchurch outlet fitting for 1/16 in.
compatibility
for use with 1/16 in. O.D. tubing
General description
PEEK housing with in-line filter connects capillary tubing (using Microtight tubing sleeves) or 1/16 in. O.D. tubing to female 10-32 fitting. Swept volume of 0.3 μL. Includes 5 × 0.5 μm frits (one installed).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2