Skip to Content
Merck
All Photos(1)

Key Documents

48980

Supelco

4,4′-DDT solution

200 μg/mL in isooctane, PESTANAL®, analytical standard

Synonym(s):

1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane solution, 4,4′-DDT solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl5
CAS Number:
Molecular Weight:
354.49
Beilstein:
1882657
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 10 mL

concentration

200 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

room temp

SMILES string

Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl

InChI

1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H

InChI key

YVGGHNCTFXOJCH-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-DDT is classified under the group of organochlorine pesticides (OCPs) belonging to the group of persistent organic pollutants (POPs).

Application

4,4′-DDT may be used as an analytical reference standard for the determination of 4,4′-DDT in:
  • Air particulate matter by ultrasound assisted extraction micro-scale cell (UAE-MSC) in conjunction with gas chromatography-mass spectrometry operating in negative chemical ionization (GC-MS-NCI).
  • Cat hair samples by two dimensional gas chromatography (GC x GC) combined with high resolution time-of-flight mass spectrometry (HR-TOFMS).
  • Water samples by thermal desorption (TD)-GC combined with full scan MS as well as liquid injection-GC combined with MS operating in selected ion monitoring (SIM) detection.
  • Wildlife liver and serum by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and GC coupled to tandem quadrupole mass spectrometry (GC-MS/MS) with MRM detection.
  • Marine pore water by solid-phase microextraction (SPME) and TD-GC-MS with SIM detection.
  • Surface soil and raw food samples by GC equipped with electron capture detector (ECD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Residues of organochlorine pesticides in surface soil and raw foods from rural areas of the Republic of Tajikistan.
Barron MG, et al.
Environmental Pollution (Barking, Essex : 1987), 224(6), 494-502 (2017)
Determination of 1-chloro-4-[2, 2, 2-trichloro-1-(4-chlorophenyl) ethyl] benzene and related compounds in marine pore water by automated thermal desorption-gas chromatography/mass spectrometry using disposable optical fiber.
Eganhouse RP and DiFilippo EL
Journal of Chromatography A, 1415(6), 38-47 (2015)
Comprehensive two-dimensional gas chromatography coupled to high resolution time-of-flight mass spectrometry for screening of organohalogenated compounds in cat hair
Brits M, et al.
Journal of Chromatography A, 1536, 151-162 (2018)
E Beristain-Montiel et al.
Journal of chromatography. A, 1477, 100-107 (2016-11-30)
New clean technologies are needed to determine concentration of organic pollutants without generating more pollution. A method to extract Persistent Organic Pollutants (POPs) from airborne particulate matter was developed using a novel technology recently patented called ultrasound assisted extraction micro-scale
Determination of organochlorine pesticides in wildlife liver and serum using gas chromatography tandem quadrupole mass spectrometry.
Bokhart M, et al.
Journal of Chromatography and Separation Techniques, 6(6), 1-1 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service