Skip to Content
Merck
All Photos(1)

Documents

40993

Supelco

1,2-Dibromo-3-chloropropane solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1,2-Dibromo-3-chloropropane.
Report on carcinogens : carcinogen profiles, 10, 79-80 (2004-08-25)
Jinwook Chung et al.
Biodegradation, 18(2), 199-209 (2006-07-06)
We tested the hypothesis that the H(2)-based membrane biofilm reactor (MBfR) is capable of reducing multiple oxidized contaminants, a common situation for groundwater contamination. We conducted bench-scale experiments with three groundwater samples collected from California's San Joaquin Valley and on
[Evidence of toxic effect of a chemical on reproduction ignored].
Maria Albin et al.
Lakartidningen, 106(37), 2296-2297 (2009-10-13)
Jonathan Hofmann et al.
International journal of occupational and environmental health, 12(4), 321-328 (2006-12-16)
The nematocide 1,2-dibromo-3-chloropropane (DBCP), widely used in Costa Rica during the late 1960s and 1970s, causes sterility in men and is a possible carcinogen. Mortality among a cohort of Costa Rican banana plantation workers was investigated. The cohort included 40,959
Paternal exposures: altered sex ratios.
J Trasler
Teratology, 62(2), 77-78 (2000-08-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service