Skip to Content
Merck
All Photos(1)

Documents

38117

Supelco

Mepiquat chloride

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,1-Dimethylpiperidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16ClN
CAS Number:
Molecular Weight:
149.66
Beilstein:
3914089
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

[Cl-].C[N+]1(C)CCCCC1

InChI

1S/C7H16N.ClH/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1

InChI key

VHOVSQVSAAQANU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Mepiquat chloride is a synthetic plant growth regulator that is absorbed by the foliage and translocated throughout the plant. It is widely used to manipulate plant structure and helps to combat excessive vegetative growth, which if left uncontrolled can lead to undesirable fruit shed, boll rot, and yield reductions. Mepiquat chloride inhibits gibberellic acid synthesis, thus reducing cell enlargement and cell division rate. Application of mepiquat chloride in cotton results in a more compact plant caused by shortened internode elongation, reduced main stem nodes and decreased leaf expansion and leaf area.

Mepiquat chloride is approved for use in European Union (EU) following Regulation (EC) No 1107/2009 (repealing Directive 91/414/EEC). The maximum residue level (MRL) for Mepiquat (sum of mepiquat and its salts, expressed as mepiquat chloride) is established from 0.02 mg/kg to 4 mg/kg for many plant-based products and 40 mg/kg for linseed, mustard, sunflower, and poppy seeds. For products of animal origin, the MRL is set from 0.05 mg/kg to 0.8 mg/kg as per Reg. (EU) 2021/976.

Mepiquat has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Mepiquat chloride CRM can also be used to: 
  • Evaluate the influence of plant density on cotton response to mepiquat chloride application in order to isolate a specific combination for optimizing plant stature and yield
  • Quantify the growth and physiological responses of cotton plants to temperature regimes and mepiquat chloride rates
  • Quantify the efficacy of mepiquat chloride on plant height, culm morphological characteristics, lignin accumulation, related enzyme activities, and its relationship with lodging resistance in maize
  • Evaluate the roles of gibberellic acid metabolism and signaling in mepiquat chloride-induced inhibition of internode elongation in cotton seedlings
  • Evaluate the effects of different mepiquat chloride concentrations on morphological and physiological changes in Eucalyptus clones

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service