29432-U
Certified screw thread vials, 10-425 thread, 12 x 32 mm
volume 2 mL, clear glass vial, PTFE/silicone septum, (convenience kit), pkg of 100 ea
All Photos(1)
About This Item
Recommended Products
material
PTFE/silicone septum
clear glass vial
polypropylene cap
feature
closure type screw top vial
kit with vial, cap, and septa
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 6 mm
volume
2 mL
fitting
thread for 10-425
Related Categories
Other Notes
The inside diameter of vials vary with the manufacture. The following insert part numbers are compatible with these large opening vials.
- 29435-UPK100, Certified 200 μL glass inserts, mandrel precision point, 6 X 29mm, with bottom spring
- 29436-UPK100, Certified 250 μL glass insert, conical bottom, 6 x 31 mm
- 29437-UPK100, Certified 300 μL glass inserts, conical bottom, 6 x 30 mm
- 29441-U PK100, Certified 350 μL glass inserts, flat bottom, 6 x 31mm
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)