27304-U
Headspace vials convenience kit (clear glass, bevel top, round bottom), pkg of 100
volume 10 mL, vial O.D. × H × I.D. 23 mm × 46 mm × 12.5 mm, PTFE/butyl rubber septum
About This Item
material
PTFE/butyl rubber septum
clear glass vial
silver aluminum seal (pressure release)
feature
closure type crimp seal
closure type crimp top vial
vial type headspace
packaging
pkg of 100 ea
vial O.D. × H × I.D.
23 mm × 46 mm × 12.5 mm
volume
10 mL
Looking for similar products? Visit Product Comparison Guide
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service