Skip to Content
Merck
All Photos(1)

Documents

20984

Supelco

Tenax® TA

stainless steel TD tube, O.D. × I.D. × L 1/2 in. × 12 mm × 7 in., preconditioned, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

material

stainless steel TD tube

Agency

EPA TO-1,IP-1B

description

Sealed with Glass Storage Container

feature

preconditioned

packaging

pkg of 1 ea

O.D. × I.D. × L

1/2 in. × 12 mm × 7 in.

matrix

Tenax TA porous polymer

application(s)

environmental
industrial hygiene

compatibility

for use with Tekmar® AEROTrap 6000 models

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tekmar is a registered trademark of Tekmar Co.
Tenax is a registered trademark of Buchem B.V.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service