Skip to Content
Merck
All Photos(1)

Key Documents

15101AST

Supelco

Astec® CHIROBIOTIC® V2 Chiral HPLC Guard Column

Column, 5 μm particle size, L × I.D. 2 cm × 1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel column

Agency

suitable for USP L88

description

HPLC guard column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 1 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

vancomycin phase

particle size

5 μm

pore size

200 Å

operating pH range

3.5-7.0

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Malakova et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3226-3230 (2009-08-01)
Warfarin is a well-known anticoagulant agent that occurs in two enantiomers, (R)-(+)-warfarin and (S)-(-)-warfarin. A new liquid chromatography method for the determination of both enantiomers was developed, validated and applied in in vitro studies with the aim of evaluating the
Chenghao Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1011, 45-52 (2016-01-12)
A chiral UFLC-MS/MS method was established and validated for quantifying d-threo-methylphenidate (d-threo-MPH), l-threo-methylphenidate (l-threo-MPH), d-threo-ethylphenidate (d-threo-EPH), l-threo-ethylphenidate (l-threo-EPH) and d,l-threo-ritalinic acid (d,l-threo-RA) in rat plasma over the linearity range of 1-500ng/mL. Chiral separation was performed on an Astec Chirobiotic V2
László Sipos et al.
Journal of chromatography. A, 1232, 142-151 (2011-12-20)
The enantiomers of four unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on chiral stationary phases containing macrocyclic glycopeptide antibiotics teicoplanin (Astec Chirobiotic T and T2), teicoplanin aglycone (Chirobiotic TAG), vancomycin (Chirobiotic V) and vancomycin aglycone (Chirobiotic VAG) as chiral selectors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service