Skip to Content
Merck
All Photos(1)

Key Documents

150711

Supelco

Titriplex® III solution

0.1 M, for 3S Adapter technology, set incl. Absorber cartridge with soda lime, for 3S adapter technology

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt solution, (Ethylenedinitrilo)tetraacetic acid disodium salt, EDTA disodium salt, EDTA-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N(CH2CO2Na)CH2CO2H]2
CAS Number:
Molecular Weight:
336.21
Beilstein:
3822669
MDL number:
UNSPSC Code:
12352107

Product Name

Titriplex® III solution, 0.1 M, for 3S Adapter technology, set incl. Absorber cartridge with soda lime, for 3S adapter technology

form

liquid

reaction suitability

reaction type: Complexometric reactions

concentration

0.1 M

technique(s)

titration: suitable

SMILES string

[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

InChI key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Contains 1506941000 (Titriplex III solution 0.1 M) and 1507040001 (cartridge filled with soda lime). Before opening the 3S cap, insert cartridge into cap.

Please see here SigmaAldrich.com/3S

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service