Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2388

Sigma-Aldrich

Anti-Histone H3.2 Antibody, clone 1K5-L1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-GFAP Antibody, clone GA5 ZooMAb® Mouse Monoclonal, H3-clustered histone 13, H3-clustered histone 14, H3-clustered histone 15, Histone H3/m, Histone H3/o

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1K5-L1, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.39 kDa
observed mol wt ~15 kDa

purified by

using Protein A

species reactivity

human, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1K5-L1 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Histone H3.2. It targets an epitope within 12 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal half of human Histone H3.2.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat2 cell acid extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H3.2 in acid extract of Rat2 cells.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H3.2 in K562 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Histone H3.2 in Human small intestine tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Histone H3.2 peptide with a KD of 4.3 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Histone H3.2 (UniProt: Q71DI3; also known as H3-clustered histone 13, H3-clustered histone 14, H3-clustered histone 15, Histone H3/m, Histone H3/o) is encoded by the HIST2H3A (also known as H3C15, H3C14, H3F2, H3FM, HIT2H3C, HIST2H3D, H3C13) gene (Gene ID: 126961) in human. Histones are highly conserved basic nuclear proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones known as histone code, and nucleosome remodeling. Mammalian cells express histone H3 variants: H3.1, H3.2, and H3.3, which show different genomic localization pattern. Histone H3.1 serves as the canonical histone, which is incorporated during DNA replication and is only expressed in mammals. Histone H3.2 is found in all eukaryotes except for yeast. It differs

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service