Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2228

Sigma-Aldrich

Anti-HIF-2α/EPAS1 Antibody, clone 2L21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-GATA4 Antibody, clone 1K15 ZooMAb® Rabbit Monoclonal, Basic-helix-loop-helix-PAS protein MOP2, Class E basic helix-loop-helix protein 73, EPAS-1, Endothelial PAS domain-containing protein 1, HIF-1-alpha-like factor, HIF-2-alpha, HIF2-alpha, HLF, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, PAS domain-containing protein 2, bHLHe73

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2L21, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 96 kDa
observed mol wt ~150 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... EPAS1(2034)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2L21 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects HIF-2 / EPAS1. It targets an epitope within 22 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human HIF-2 /EPAS1.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HIF-2 /EPAS1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected His/ABP-tagged recombinant fragment of Human HIF-2 /EPAS1.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HIF-2 /EPAS1 in HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound HIF-2 /EPAS1 peptide with a KD of 1.1 x 10-8 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: Serial of dilutions from a representative lot detected His/ABP-tagged recombinant fragment of Human HIF-2 /EPAS1.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Endothelial PAS domain-containing protein 1 (UniProt: Q99814; also known as EPAS-1, Basic-helix-loop-helix-PAS protein MOP2, Class E basic helix-loop-helix protein 73, bHLHe73, HIF-1-alpha-like factor, HLF, Hypoxia-inducible factor 2-alpha, HIF-2-alpha, HIF2-alpha, Member of PAS protein 2, PAS domain-containing protein 2) is encoded by the EPAS1 (also known as BHLHE73, HIF2A, MOP2, PASD2) gene (Gene ID: 2034) in human. HIF-2 is a transcription factor that is involved in the induction of oxygen regulated genes. It displays about 48% identity to HIF-1 and is widely expressed with high expression observed in endothelial cells, placenta, lung, and heart. Its levels are strongly induced by hypoxic conditions. HIF-2 dimerizes with ARNT, via amino acid sequence 171-192, and this heterodimer binds to core DNA sequence 5′-TACGTG-3′ within the hypoxia response element (HRE) of target gene promoters. Its DNA-binding region is localized to amino acids 26-53. It has two PAS (Per-Arnt-Sim) dom

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service