ZRB1885
Anti-CHD-8 Antibody, clone 1H9 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Chromodomain-helicase-DNA-binding protein 8;EC: 3.6.4.12;ATP-dependent helicase CHD8;Helicase with SNF2
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1H9, recombinant monoclonal
description
1H9 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 290.52 kDa
observed mol wt ~290 kDa
purified by
using Protein A
species reactivity
mouse, human
species reactivity (predicted by homology)
monkey
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
Internal
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CHD8(57680)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in EL4 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CHD-8 in EL4 cell lysate.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CHD-8 in nuclear extracts from NIH3T3 and HeLa cells.
Affinity Binding Assay: A representative lot of this antibody bound CHD-8 peptide with a KD of 5.1 x 10-6 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CHD-8 in human prostate tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western Blotting in EL4 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CHD-8 in EL4 cell lysate.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service