Skip to Content
Merck
All Photos(16)

Documents

ZRB1759

Sigma-Aldrich

Anti-CD4 Antibody, clone 2G19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G19, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~51.11 kDa
observed mol wt ~51 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2G19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects CD4. It targets an epitope within 18 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal, extracellular domain of human CD4.

Application

Quality Control Testing

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD4 in HL-60 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD4 in THP-1 cell lysate.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected CD4 in one million THP-1 cells.

Affinity Binding Assay: A representative lot of this antibody bound CD4 peptides with a KD of 4.5 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD4 in human spleen and human colon tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD4 in THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD4, clone 2G19 ZooMAb®, Cat. No. ZRB1759, is a recombinant Rabbit monoclonal antibody that targets CD4 and has been tested in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

T-cell surface glycoprotein CD4 (UniProt: P01730; also known as T-cell surface antigen T4/Leu-3, CD4) is encoded by the CD4 gene (Gene ID: 920) in human. CD4 is a single-pass type I membrane glycoprotein that is highly expressed in T-helper cells and is also found in macrophages, dendritic cells, and NK cells. CD4 is synthesized with a signal peptide (a 1-25), which is subsequently cleaved off to generate the mature form that has an extracellular domain (aa 26-396), a transmembrane domain (aa 397-418), and a cytoplasmic domain (aa 419-458). It has three Ig-like C2-type and one Ig-like V-type domain. CD4 plays an essential role in the immune response and serves multiple functions in response against both external and internal offenses. In T-cells, it functions primarily as a coreceptor for MHC class II molecule:peptide complex. It interacts simultaneously with the T-cell receptor (TCR) and the MHC class II presented by antigen presenting cells and recruits the Src kinase LCK to the vicinity of the TCR-CD3 complex. The LCK can then initiate different intracellular signaling events by phosphorylating various substrates, which lead to lymphokine production, motility, adhesion and activation of T-helper cells. CD4 is also reported to participates in the development of T-helper cells in the thymus and trigger the differentiation of monocytes into functional mature macrophages. CD4 also serves a primary receptor for human immunodeficiency virus-1 (HIV-1). It is removed from plasma membrane by Nef protein of HIV-1, which increases clathrin-dependent endocytosis and lysosomal degradation of CD4. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service