Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1034

Sigma-Aldrich

Anti-ULBP-4/RAET1E Antibody, clone 6C10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Lymphocyte effector toxicity activation ligand, N2DL-4, NKG2D ligand 4, NKG2DL4, RAE-1-like transcript 4, Retinoic acid early transcript 1E, UL16-binding protein 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€163.00
4 X 25 μL
€335.00

€163.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μL
€163.00
4 X 25 μL
€335.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€163.00


In StockDetails


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6C10, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.12 kDa
observed mol wt ~26 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RAET1E(135250)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 6C10 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Retinoic acid early transcript 1E. It targets an epitope within the extracellular domain.

Immunogen

His-tagged recombinant fragment corresponding to 195 amino acids from the extracellular domain of human Retinoic acid early transcript 1E (RAET1E).

Application

Anti-ULBP-4/RAET1E, clone 6C10 ZooMAb, Cat. No. ZRB1034, is a recombinant Rabbit monoclonal antibody that specifically targets Retinoic acid early transcript 1E and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ULBP-4/RAET1E in ULBP-4 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ULBP-4/RAET1E in HT-29 cells.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant ULBP-4 protein with a KD of 4.5 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Retinoic acid early transcript 1E (UniProt: Q8TD07; also known as Lymphocyte effector toxicity activation ligand, NKG2D ligand 4, N2DL-4, NKG2DL4, RAE-1-like transcript 4, UL16-binding protein 4, ULBP-4) is encoded by the RAET1E (also known as LETAL, N2DL4, ULBP4, UNQ1867/PRO4303) gene (Gene ID: 135250) in human. ULBP-4 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-30), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 31-225), a transmembrane domain (aa 226-248), and a cytoplasmic domain (aa 249-263). It is predominantly expressed in the skin, but also expressed in testis and trachea. Its levels are upregulated in tumor cells of different origins and its expression progressively decreased after treatment of tumor cells with retinoic acid. ULBP-4 also serves as a functional ligand for NKG2D. ULBP-4 is reported to bind and co-stimulate NKG2D-expressing effector cells including NK cells, NKT cells, CD8+ T cells. ULBP-4 has a MHC class I alpha-1 like (aa 31-116) and MHC class I alpha-2 like (117-207) regions that down-regulate the cell surface expression of Killer Cell Lectin Like Receptor K1 (KLRK1). Six different isoforms of ULBP-4 have been described that are produced by alternative splicing. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chalupny, NJ., (2003). Biochem. Biophys. Res. Commun. 305(1); 129-135).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service