Key Documents
Z741028
Nalgene® media bottles, style 2020
capacity 2000 mL
Synonym(s):
bottle nalgene, nalgene containers, sterile bottles
Select a Size
12 EA
€336.00
€336.00
In StockDetails
Select a Size
Change View
12 EA
€336.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
€336.00
In StockDetails
material
high-density polyethylene cap
square bottle
transparent PETG
Joint
Joints threaded neck (53B)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no
with cap
packaging
pack of 12 ea (2 xpack of 6)
manufacturer/tradename
Nalgene® 342020-2000
capacity
2000 mL
General description
Media bottles (square). Same materials and design as Style 2019, but without shrink band around neck.
Features and Benefits
  • Extensive validation documentation is available
  • Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
  • Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
  • Non-pyrogenic and non-cytotoxic
  • Animal derived component free (ADCF) material for bottle and closure material.
  • Leakproof and transparent
  • Devoid of costly washing, depyrogenation steps
  • Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)