Skip to Content
Merck
All Photos(1)

Documents

Z717436

Syringe tips for Nichiryo 8100 repetitive volume dispenser

without tip, maximum volume 0.06 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

sterility

non-sterile

feature

electronic: no
single-channel

packaging

pack of 100 ea (syringe (0.06 mL))

manufacturer/tradename

Nichiryo SG-V2

availability

not available in (Not available in France)

maximum volume

0.06 mL

Looking for similar products? Visit Product Comparison Guide

General description

Syringes are autoclavable for re-use, saving plastic waste and cost (except 3 mL and 60 mL syringes).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service