Documents
Z693715
Duran® filtering flask, with Keck® assembly set
capacity 1000 mL
Synonym(s):
212045458
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
Joint
joint ST/NS neck45 mm I.D.
sterility
non-sterile
packaging
pkg of 1 ea
pkg of 10 ea
manufacturer/tradename
DURAN® 212045458
availability
not available in USA and Canada
capacity
1000 mL
General description
The Duran filtering flask includes a Keck clip assembly and is idea for vacuum filtrations. The plastic hose connections meet safety and product regulations, and the flask has the Erlenmeyer shape.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Keck is a registered trademark of Adcuram Lab Technologies GmbH