Key Documents
Z692034
3DKUBE microscope stage adapter, universal
L × W × H 1 in. × 1 in. × 3 in.
Synonym(s):
3D, 3D Cell Culture
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea
manufacturer/tradename
Kiyatec AC010
L × W × H
1 in. × 1 in. × 3 in.
General description
The Universal Microscope Stage Adapter provides a convenient and universal adapter to affix 3DKUBE 3D Cell Culture Plasticware into common microscopy platforms to accommodate in situ imaging via the plasticware′s optically clear imaging window. Stage adapter has the same footprint as a conventional microscope slide.
Legal Information
3DKUBE is a trademark of Kiyatec, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Karolina Janeczek Portalska et al.
Journal of tissue engineering and regenerative medicine, 10(5), 363-373 (2013-04-18)
Modular tissue engineering is a strategy to create scalable, self-assembling, three-dimensional (3D) tissue constructs. This strategy was used to deliver endothelial-like cells derived from bone marrow mesenchymal stromal cells (EL-MSCs) to locally induce vascularization. First, tissue engineered modules were formed
Sayed-Hadi Mirmalek-Sani et al.
Biomaterials, 34(22), 5488-5495 (2013-04-16)
Emergent technologies of regenerative medicine have the potential to overcome the limitations of organ transplantation by supplying tissues and organs bioengineered in the laboratory. Pancreas bioengineering requires a scaffold that approximates the biochemical, spatial and vascular relationships of the native