Z654353
Consort EV series power supplies
EV231, output0-300 V, 0-1,000 mA, 0-150 W
Synonym(s):
Consort EV series power supplies
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
AC/DC input
230 V AC
description
10 ohm min load resistance
4 × 4 mm sockets
manufacturer/tradename
EV231
availability
available only in EU
parameter
0-40 °C ambient temperature
W × D × H
24 cm × 20 cm × 13 cm
weight
4.3 kg
output
0-300 V, 0-1,000 mA, 0-150 W
General description
- Timer 0-99.59 hour
- Volt hours 0-99.99 kVH
- LCD display, 2 × 16 characters
- Ground leakage detection 500 μA
- No load detection, programmable, overload detection
- 3600 values of data logging
- 0-95% non condensing humidity
All of the Consort EV series power supplies offer manual programming of voltage, current, temporary changes to power and time parameters can be made without interrupting the run. Up to 9 frequently used programs can be stored for future recall.
The instruments are fully protected against overload condition including accidental short circuit of output. Galvanic RS232 input/output insulation prevents ground loop interferences when connected to computer.
The instruments are fully protected against overload condition including accidental short circuit of output. Galvanic RS232 input/output insulation prevents ground loop interferences when connected to computer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service