Key Documents
Z368997
Finnpipette® stand
pkg of 1 ea
Select a Size
1 EA
€95.30
€95.30
Check Cart for Availability
Select a Size
Change View
1 EA
€95.30
About This Item
UNSPSC Code:
41122801
NACRES:
NB.01
€95.30
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Finn 9420290
Finn
General description
Rugged molded supports provide plenty of height for the longest Finnpipettes. Aluminum crossbar has curved upper edge to fit securely under the finger support of any Finnpipette single-channel or multi-channel pipette.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)