Z363413
Scienceware® gel fixing tray
L × W × H 16 in. (40 cm) × 20 in. (50 cm) × 3 in. (7 cm)
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
L × W × H
16 in. (40 cm) × 20 in. (50 cm) × 3 in. (7 cm)
General description
Constructed of heavy polyethylene with a stiffening flange at the top and ridges in the bottom to prevent suctioning of glass plates. Chemically resistant to most common staining and fixing mixtures. The drain stopcock in the side with 1.8 meters (6 ft.) Vikem® vinyl tubing 13 mm (1/2 in.) I.D. allows more complete drainage and a more controlled drain rate to avoid disturbing the gel. Hooks on the rim of the tray hold the tubing. Inside dimensions given.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)