Z326453
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
volume 200 mL, accuracy: 0.15 mL
Synonym(s):
Flasks
Select a Size
€81.60
Available to ship on01 April 2025Details
Select a Size
About This Item
€81.60
Available to ship on01 April 2025Details
Recommended Products
material
borosilicate glass 3.3 flask
Joint
joint ST/NS neck14/23
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37408
volume measuring range
200 mL, accuracy: 0.15 mL
capacity
200 mL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service