Skip to Content
Merck
All Photos(1)

Documents

Z315613

Eppendorf® Varispenser® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

description

precision ± 0.1%
provided with adapters for 28, 40 and 45 mm threads.
telescopic intake tube allows use with short or tall bottles.

feature

fits 32 mm bottle thread

manufacturer/tradename

Eppendorf® 4961 000.012

volume measuring range

0.5-2.5 mL, accuracy: 0.6%, resolution: 0.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Bottle-top dispensers with PTFE valve blocks and pistons and a Pt-lr spring bolosilicate ball valve for high resistance to aggressive chemicals. Modular construction for simple cleaning and maintenance.

Safety valve for priming without loss of reagent and fine adjustment of volume.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service