Skip to Content
Merck
All Photos(1)

Key Documents

TTR010

Sigma-Aldrich

Reticulum TISSUE-TROL Control Slides

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€255.00

€255.00


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 EA
€255.00

About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

€255.00


Available to ship on08 April 2025Details


Request a Bulk Order

biological source

human liver (tissue containing reticular fibers)

application(s)

hematology
histology

storage temp.

room temp

Application

Histology slides with paraffin embedded section of human liver tissue containing reticular fibers. Suitable for use with Reticulum Stain Procedure No. HT102.

Components

Box contains 24 unstained slides plus one reference slide stained using Reticulum Stain Procedure No. HT102.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service