Skip to Content
Merck
All Photos(2)

Key Documents

T7540

Sigma-Aldrich

Tyrphostin 47

≥98%

Synonym(s):

α-Cyano-3,4-dihydroxythiocinnamamide, 3,4-Dihydroxy-α-cyanothiocinnamamide, RG 50864

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O2S
CAS Number:
Molecular Weight:
220.25
Beilstein:
6508337
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

potency

2.4 μM IC50

solubility

DMSO: soluble 50 mg/mL, clear, orange
ethanol: soluble

storage temp.

2-8°C

SMILES string

NC(=S)\C(=C\c1ccc(O)c(O)c1)C#N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Tyrphostin 47 has been used in LOPAC screening assays to identify human tissue transglutaminase (TGM2) inhibitors. Tyrphostin 47 has also been used to study its effects on hyperpolarization-activated cation current in porcine pacemaker cells.

Biochem/physiol Actions

Tyrphostin 23 is a small molecule inhibitor of epidermal growth factor (EGF) receptor kinase function. This molecule associates with the substrate subsite of the protein tyrosine kinase (PTK) domain,.
EGFR tyrosine kinase inhibitor.

Preparation Note

Tyrphostin 47 is is soluble in DMSO (50 mg/ml) and yields a clear, orange solution. The product is also soluble in ethanol (40 mM).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shibata et al.
British journal of pharmacology, 128(6), 1284-1290 (1999-12-01)
1 The hyperpolarization-activated cation current (If) was recorded in single pacemaker cells of the porcine sino-atrial node, and the effects of genistein, an isoflavone inhibitor of tyrosine-specific protein kinases was investigated by the whole-cell patch clamp technique. 2 Genistein (20-500
P L Lorenzo et al.
Equine veterinary journal, 34(4), 378-382 (2002-07-16)
Epidermal growth factor (EGF) has been shown to have a positive effect during oocyte in vitro maturation in several species. This study was performed to establish the capacity of equine oocytes to undergo nuclear maturation in the presence of EGF
G Kéri et al.
Annals of the New York Academy of Sciences, 1010, 109-112 (2004-03-23)
Selective inhibition of the "false" proliferative signals via targeting tyrosine kinases resulting in the induction of apoptosis by depletion of the "survival factors" is one of the most studied and widely accepted concepts of modern chemotherapy. We have synthesized a
Charles W Leffler et al.
American journal of physiology. Heart and circulatory physiology, 285(1), H292-H297 (2003-03-08)
Carbon monoxide (CO) is produced from heme by heme oxygenase-2 (HO-2) in cerebral blood vessels. Gas chromatography-mass spectrometry was used on piglet cerebral microvessels to address the hypothesis that CO production is regulated by heme delivery and HO-2 catalytic activity.
J Thundathil et al.
Molecular human reproduction, 8(9), 811-816 (2002-08-30)
Human sperm capacitation involves complex signal transduction mechanisms during which double phosphorylation of the threonine-glutamine-tyrosine motif (P-Thr-Glu-Tyr-P) occurs in some sperm proteins. The objective of this study was to investigate the regulation of this process. Fetal cord serum ultrafiltrate (FCSu)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service