T3524
4-(2-Thiazolylazo)resorcinol
Synonym(s):
2-(2,4-Dihydroxyphenylazo)thiazole, TAR
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H7N3O2S
CAS Number:
Molecular Weight:
221.24
Beilstein:
204257
EC Number:
MDL number:
UNSPSC Code:
12171500
mp
211 °C (dec.) (lit.)
SMILES string
Oc1ccc(\N=N\c2nccs2)c(O)c1
InChI
1S/C9H7N3O2S/c13-6-1-2-7(8(14)5-6)11-12-9-10-3-4-15-9/h1-5,13-14H/b12-11+
InChI key
SHNIKUXMZFPPCS-VAWYXSNFSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service