Skip to Content
Merck
All Photos(1)

Key Documents

T2896

Sigma-Aldrich

Triflupromazine hydrochloride

Synonym(s):

N,N-Dimethyl-2-(trifluoromethyl)-10H-phenothiazine-10-propanamine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19F3N2S · HCl
CAS Number:
Molecular Weight:
388.88
Beilstein:
3801519
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Cl.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(F)(F)F

InChI

1S/C18H19F3N2S.ClH/c1-22(2)10-5-11-23-14-6-3-4-7-16(14)24-17-9-8-13(12-15(17)23)18(19,20)21;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H

InChI key

FTNWXGFYRHWUKG-UHFFFAOYSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

Application

Triflupromazine hydrochloride has been used for the screening of inhibitors of S. mansoni miracidial transformation5. It has also been used to study its intracellular localization in central nervous system cells using secondary ion mass spectrometry microscopy6.

Biochem/physiol Actions

Triflupromazine is a D2 dopamine antagonist and a phenothiazine antipsychotic. It is also a potent tranquilizer7 and can significantly reduce body temperature8.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L L Zhang et al.
Biology of the cell, 74(1), 99-103 (1992-01-01)
The intracellular localization of two neurotropic drugs, flunitrazepam (benzodiazepine) and triflupromazine (phenothiazine), was studied by secondary ion mass spectrometry microscopy (SIMS) in three varieties of cells. The images of the intracellular distributions of the two drugs are easily obtained by
A study of triflupromazine in dominant-lethal, cytogenetic and host-mediated assays.
V A Ray et al.
Mutation research, 18(3), 301-309 (1973-06-01)
D Jovanović-Mićić et al.
European journal of pharmacology, 272(1), 21-30 (1995-01-05)
Intracerebroventricular injection of dopamine (0.5-4.0 mg) produced dose-dependent and short-lasting emesis (1-8 min) in cats, which was abolished after ablation of the area postrema. Relatively selective alpha 2-adrenoceptor antagonists (yohimbine and idazoxan) and a mixed alpha 1- and alpha 2-adrenoceptor
The structure of triflupromazine, a phenothiazine 'tranquilizer' drug molecule.
Phelps, D. W., and Cordes, A. W.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 30(12), 2812-2816 (1974)
M Tatsumi et al.
European journal of pharmacology, 368(2-3), 277-283 (1999-04-08)
Using radioligand binding techniques, we determined the equilibrium dissociation constants (K(D)) for 37 neuroleptics and one metabolite of a neuroleptic (haloperidol metabolite) for the human serotonin, norepinephrine, and dopamine transporters with [3H]imipramine, [3H]nisoxetine, and [3H]WIN35428, respectively. Among neuroleptics, the four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service