Skip to Content
Merck
All Photos(1)

Documents

SYNB135430

Synthware vacuum desiccator

includes a ceramic plate and lid with stopcock/port, ID 300mm

Synonym(s):

Synthware B135430, desiccator with Top Knob

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

packaging

pack of 1 ea

manufacturer/tradename

Kemtech B135430

diam.

300 mm

Looking for similar products? Visit Product Comparison Guide

General description

Vacuum Desiccator, ID 300mm, Includes Lid & Ceramic Plate includes Accessories: A682440G

Application

kemetch; synthware; vacuum desiccator; desiccator

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service