SRP6278
TDRD12-BD2 (908-999) his tag human
recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Synonym(s):
ECAT8
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (SDS-PAGE)
form
liquid
mol wt
13 kDa
packaging
pkg of 1 mg
pkg of 100 μg
pkg of 20 μg
concentration
2 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... TDRD12(91646)
General description
Tudor domains are small protein structural motifs of about ~50 amino acids related to the “royal family” of methyl readers, which also includes chromo, MBT, PWWP, and Agenet-like domains. Tudor domains occur either alone, in tandem, or with other domains and are found in many proteins that are involved in RNA metabolism, germ cell development, transposon silencing, DNA damage response, histone modification, and chromatin remodeling. The tudor domains recognize symmetric methylated arginine or methylated lysine residues. Tudor domain proteins act as an oncogene and play a very important role in HCC and colon cancer. TDRD is also involved in RISC complex and interacts with AEG-1 oncogene. The tudor domain can bind to methylated arginine protein and promote tumor angiogenesis in human hepatocellular carcinoma, etc. The recombinant protein includes TDRD12 (domain 908-999aa) with N-terminal His-tag.
Physical form
rh-TDRD12-BD2 is supplied as a solution in PBS Buffer containing 10% glycerol.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service