Skip to Content
Merck
All Photos(1)

Key Documents

SRP6174

Sigma-Aldrich

Ubiquitin-K0 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), 95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)
95% (HPLC)

form

liquid

mol wt

8.760 kDa

packaging

pkg of 500 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RPS27A(6233)

General description

Ubiquitin is a small polypeptide that can be conjugated via its C-terminus to amine groups of lysine residue on target proteins. This conjunction is referred to as monoubiquitylation. Additional ubiquitin moieties can be subsequently conjugated to this initial ubiquitin, utilizing any one of the seven lysine residues on the surface of ubiquitin. The formation of these ubiquitin chains is referred to as polyubiquitylation. This tag-free recombinant form of human ubiquitin is engineered to have all available lysines mutated to arginines. This molecule will not participate in polyubiquitin formation, and can therefore be used in control experiments for the study of ubiquitylation. Covalent attachment of ubiquitin to other proteins serves various functions, but its major role is to target cellular proteins for destruction. Cellular components that activate, transfer, remove, or simply recognize ubiquitin number in the hundreds, perhaps even in the thousands. In light of this complexity the ubiquitin pathway is ideal for a systems biology approach. Ubiquitin plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

Physical form

4 mg/mL in PBS.

Preparation Note

Centrifuge the vial prior to opening.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service