Skip to Content
Merck
All Photos(1)

Key Documents

SRP5346

Sigma-Aldrich

CyclinC, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCNC, CycC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~55 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CCNC(892)

General description

Cyclin C is a member of the cyclin family of proteins. The gene encoding it is localized on human chromosome 6q16.2.

Biochem/physiol Actions

Cyclin C interacts with the cyclin-dependent kinase 8 (CDK8) and induces the phosphorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II. The mRNA levels of Cyclin C fluctuate during the cell cycle and peak at the G1 phase. Cyclin C acts as a critical regulator of the G0/G1 transition of human hematopoietic stem cells. Cyclin C can also associate with CDK3 to enhance retinoblastoma phosphorylation at S807/811 during the G0/G1 transition, and this phosphorylation is required for cells to exit G0 efficiently.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclin C/cdk3 promotes Rb-dependent G0 exit.
Ren S and Rollins BJ
Cell (2004)
Genome-wide analysis of genetic
alterations in acute lymphoblastic
leukaemia
Charles G. Mullighan
Nature (2007)
mTORC1 Down-Regulates Cyclin-Dependent Kinase 8 (CDK8) and Cyclin C (CycC)
Daorong Feng
PLoS ONE (2015)
Cyclin C Regulates Human Hematopoietic Stem/Progenitor
Cell Quiescence
YASUHIKO MIYATA
Stem Cells (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service