Skip to Content
Merck
All Photos(2)

Key Documents

SRP5252

Sigma-Aldrich

PKC-δ, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAY1, MGC49908, PRKCD, nPKC-delta

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€278.00

€278.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 μG
€278.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€278.00


In StockDetails


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

213-388 nmol/min·mg

mol wt

~104 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCD(5580)

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer Gene List
Ma Hongbao
Cancer Biology & Medicine (2016)
Identification of proteins interacting with protein kinase C-d in hyperthermia-induced apoptosis and thermotolerance of Tca8113 cells.
Liu J
Molecular Medicine Reports (2015)
Changes in nuclear protein kinase C-delta holoenzyme, its catalytic fragments, and its activity in polyomavirus-transformed pyF111 rat fibroblasts while proliferating and following exposure to apoptogenic topoisomerase-II inhibitors.
Dal Pra I
Experimental Cell Research (1999)
Loss of PKCd Induces Prostate Cancer Resistance to Paclitaxel through Activation of Wnt/?-Catenin Pathway and Mcl-1 Accumulation
M. Luz Flores
Molecular Cancer Therapeutics (2016)
Protein kinase C-d inactivation inhibits the proliferation and survival of cancer stem cells in culture and in vivo.
Chen Z
BMC Cancer (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service