Skip to Content
Merck
All Photos(1)

Key Documents

SRP5181

Sigma-Aldrich

FOS, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

c-FOS

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€470.00

€470.00


In StockDetails



Select a Size

Change View
50 μG
€470.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€470.00


In StockDetails


biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~78 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FOS(2353)

General description

FOS is a member of the FOS gene family that consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. FOS gene encodes a leucine zipper protein that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. FOS overexpression leads to decreased phosphorylation and dimerization of STAT1, which in turn downregulates p21 gene expression. This regulatory pathway may enhance the proliferation of lymphocytes in rheumatoid arthritis patients.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Saez et al.
Cell, 82(5), 721-732 (1995-09-08)
The proto-oncogene c-fos is a major nuclear target for signal transduction pathways involved in the regulation of cell growth, differentiation, and transformation. Using the multistep skin carcinogenesis model, we have directly tested the ability of c-fos-deficient mice to develop cancer.
Mari Hikasa et al.
Biochemical and biophysical research communications, 304(1), 143-147 (2003-04-23)
Features characteristic to rheumatoid arthritis (RA) including synovial overgrowth and joint destruction are experimentally produced by augmenting c-fos gene expression. We show that cyclin dependent kinase inhibitor p21waf1/cip1, that inhibits cell proliferation, is down-regulated in conjunction with up-regulation of c-fos

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service