Skip to Content
Merck
All Photos(2)

Key Documents

SRP5041

Sigma-Aldrich

JAK3 (781-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€377.00

€377.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€377.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€377.00


Estimated to ship on26 April 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

77-104 nmol/min·mg

mol wt

~64 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... JAK3(3718)

General description

JAK3 is a member of the JAK family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction. Low levels of JAK3 expression is detected in immature hematopoietic cells, which dramatically increases during terminal differentiation of these cells suggesting a role of JAK3 in the differentiation of hematopoietic cells. Mutations in JAK3 are associated with autosomal SCID (severe combined immunodeficiency disease). Mice lacking JAK3 show a severe block in B-cell development at the pre-B stage in bone marrow suggesting that JAK3 is critical for the progression of B-cell development in the bone marrow.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Russell et al.
Science (New York, N.Y.), 270(5237), 797-800 (1995-11-03)
Males with X-linked severe combined immunodeficiency (XSCID) have defects in the common cytokine receptor gamma chain (gamma c) gene that encodes a shared, essential component of the receptors of interleukin-2 (IL-2), IL-4, IL-7, IL-9, and IL-15. The Janus family tyrosine
D C Thomis et al.
Science (New York, N.Y.), 270(5237), 794-797 (1995-11-03)
Biochemical studies of signaling mediated by many cytokine and growth factor receptors have implicated members of the Jak family of tyrosine kinases in these pathways. Specifically, Jak3 has been shown to be associated with the interleukin-2 (IL-2) receptor gamma chain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service