Skip to Content
Merck
All Photos(1)

Key Documents

SRP3282

Sigma-Aldrich

SCD34 LIGAND HUMAN    

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

Hematopoietic progenitor cell antigen CD34

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€329.00

€329.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
20 μG
€329.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€329.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

human

recombinant

expressed in CHO cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

70.0-80.0 kDa

packaging

pkg of 20 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD34(947)

General description

CD34 has been widely used as a molecular marker for the identification, isolation, and manipulation of hemopoietic stem cells and progenitors. CD34 can function as a regulator of hemopoietic cell adhesion by mediating the attachment of stem cells to bone marrow stromal cells or other bone marrow components. The full length human CD34 is a 385 amino acid protein, consisting of a 31 amino acid signal sequence, a 74 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain and a 259 amino acid extracellular domain.

Biochem/physiol Actions

CD34 is a highly glycosylated type I membrane protein that is selectively expressed on hematopoietic stem cells and vascular endothelium. Recombinant human sCD34 is a 258 amino acid polypeptide containing only the extracellular domain of the full length CD34 protein.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service