SRP2009
TFIIF (RAP30+Rap74) human
recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Synonym(s):
BTF4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
recombinant
expressed in E. coli
Assay
≥80% (SDS-PAGE)
form
frozen liquid
mol wt
~84 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
400 μg/mL
color
clear colorless
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... GTF2F1(2962) , GTF2F2(2963)
Biochem/physiol Actions
The transcription factor IIF (TFIIF) is composed of 58 kDa (RAP74) and 26 kDa (RAP34) subunits as a heterodimer, and was first identified through the ability to interact with immobilized RNA polymerase II. The Rap74 subunit of TFIIF can be phosphorylated by TAF250 both in vivo and in vitro. The RAP30 subunit of TFIIF contains two distinct regions with sequence similarity to E. coli s factors and can deliver RNA polymerase II to the promoter to support transcription initiation in the absence of RAP74.
Physical form
Clear and colorless frozen liquid solution
Preparation Note
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Science (New York, N.Y.), 253(5022), 900-902 (1991-08-23)
RAP30/74 is a heteromeric general transcription initiation factor that binds to mammalian RNA polymerase II. The RAP30 subunit contains a region that is similar in amino acid sequence to the RNA polymerase-binding domain of the Escherichia coli transcription initiation factor
The Journal of biological chemistry, 260(18), 10353-10360 (1985-08-25)
We have used affinity chromatography on columns containing immobilized calf thymus RNA polymerase II to isolate three phosphoproteins (RAP72, RAP38, and RAP30) that bind directly to RNA polymerase II. All could be isolated from cell nuclei, and all three could
Nature, 341(6241), 410-414 (1989-10-05)
RAP30/74 is a heteromeric general transcription initiation factor which binds to RNA polymerase II. Here we report that preparations of RAP30/74 contain an ATP-dependent DNA helicase whose probable function is to melt the DNA at transcriptional start sites. The sequence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service