Skip to Content
Merck
All Photos(1)

Key Documents

SRP0672

Sigma-Aldrich

IDO2, His-Tag

recombinant, expressed in E. coli

Synonym(s):

3-dioxygenase 2, IDO, IDO-2, Indoleamine 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€448.00

€448.00


Check Cart for Availability


Select a Size

Change View
100 μG
€448.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€448.00


Check Cart for Availability

recombinant

expressed in E. coli

form

frozen liquid

mol wt

46 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IDO2(169355)

General description

Human IDO2, also known as Indoleamine 2,3-dioxygenase 2, GenBank Accession No. NM_194294, a.a. 15-420(end) with an N-terminal His-tag, expressed in the presence of hemin in an E. coli cell expression system. MW = 46 kDa.

Physical form

Aqueous solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service