SRP0502
CEACAM, AviTag™ ,His-Tag
recombinant, expressed in HEK 293 cells
Synonym(s):
Carcinoembryonic antigen-related cell adhesion molecule 1, biliary glycoprotein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in HEK 293 cells
form
frozen liquid
mol wt
47 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CEACAM1(634)
General description
Human CEACAM, also known as Carcinoembryonic antigen-related cell adhesion molecule 1 or biliary glycoprotein, (GenBank Accession No. NM_ 001712), amino-acids 35-428, with C-terminal Avi-His-tag, MW = 47 kDa, expressed in HEK293 cells. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.
Physical form
Aqueous solution
Legal Information
AviTag is a trademark of Avidity, LLC
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service