Skip to Content
Merck
All Photos(2)

Key Documents

SRP0457

Sigma-Aldrich

BRD3 (29-145) GST tag human

recombinant, expressed in E. coli, ≥81% (SDS-PAGE)

Synonym(s):

ORFX, RING3L, bromodomain containing 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€525.00

€525.00


Check Cart for Availability


Select a Size

Change View
100 μG
€525.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€525.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥81% (SDS-PAGE)

form

aqueous solution

mol wt

40.7 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRD3(8019)

General description

Human Bromodomain containing 3, also known as BRD3 or RING3L (GenBank Accession No. NM_007371) amino-acids 29 – 145 with N-terminal GST-tag, MW = 40.7 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Gamsjaeger et al.
Molecular and cellular biology, 31(13), 2632-2640 (2011-05-11)
Recent data demonstrate that small synthetic compounds specifically targeting bromodomain proteins can modulate the expression of cancer-related or inflammatory genes. Although these studies have focused on the ability of bromodomains to recognize acetylated histones, it is increasingly becoming clear that
Janine M Lamonica et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(22), E159-E168 (2011-05-04)
Acetylation of histones triggers association with bromodomain-containing proteins that regulate diverse chromatin-related processes. Although acetylation of transcription factors has been appreciated for some time, the mechanistic consequences are less well understood. The hematopoietic transcription factor GATA1 is acetylated at conserved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service