Skip to Content
Merck
All Photos(1)

Key Documents

SRP0399

Sigma-Aldrich

Histone H3 peptide (Mono-methylated K9), biotin

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€268.00

€268.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μL
€268.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€268.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

aqueous solution

packaging

pkg of 500 μL

concentration

40 μM

shipped in

dry ice

storage temp.

−70°C

General description

Histone H3 peptide mono-methylated at Lysine 9, biotinylated, substrate for histone demethylases belonging to the Jumanji domain-containing (JMJ) family .

Application

Suitable as a substrate for histone demethylases belonging to the Jumonji (JMJ) family. It can be used to study enzyme kinetics, and to screen small molecular inhibitors of these demethylases for drug discovery and HTS applications.

Other Notes

500 μL at 40 μM, or 20 nmol Sold as 500 μL

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiv I S Grewal et al.
Current opinion in cell biology, 16(3), 230-238 (2004-05-18)
Heterochromatin mediates various nuclear processes including centromere function, gene silencing and nuclear organization. Although it was discovered nearly 75 years ago, the pathways involved in heterochromatin establishment, assembly and epigenetic maintenance have been elusive. Recent reports have demonstrated that distinct
Judd C Rice et al.
Molecular cell, 12(6), 1591-1598 (2003-12-24)
The functional significance of mono-, di-, and trimethylation of lysine residues within histone proteins remains unclear. Antibodies developed to selectively recognize each of these methylated states at histone H3 lysine 9 (H3 Lys9) demonstrated that mono- and dimethylation localized specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service