SRP0170
ACK1 Active human
recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
Synonym(s):
Activated Cdc42-associated kinase 1, TNK2, Tyrosine kinase, non-receptor, 2
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected insect cells
Assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
68 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... TNK2(10188)
General description
ACK1 (activated CDC42 kinase 1) is a non-receptor tyrosine kinase and is present in all human tissues. It belongs to the family of cytoplasmic tyrosine kinases. The protein has a sterile α motif (SAM) domain, a tyrosine kinase domain, a SH3 (src homology 3) domain, a CDC42/RAC-interactive (CRIB) domain, and a region that is homologous to the EGFR (epidermal growth factor receptor) binding domain of Mig6 (mitogen-induced gene 6) and a ubiquitin association domain.
Human Ack1 (GenBank Accession No. NM_005781), full length (amino acids 110-476) with N-terminal GST tag, MW = 68kDa, expressed in Sf9 insect cell systems.
Human Ack1 (GenBank Accession No. NM_005781), full length (amino acids 110-476) with N-terminal GST tag, MW = 68kDa, expressed in Sf9 insect cell systems.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Biochem/physiol Actions
ACK1 (activated CDC42 kinase 1) was first discovered for its binding to CDC42 (cell division control protein 42 homolog). ACK1 and CDC42 controls cellular attachment and migration. ACK1 is involved in cell survival, proliferation and migration. It is also associated with various cancers. The ACK1 gene is overexpressed in prostate, gastric and pancreatic cancers. It also interacts with the heavy chain of clathrin and might be involved in receptor-mediated endocytosis. ACK1 is a dual specificity kinase towards WASP (Wiskott–Aldrich Syndrome protein). ACK1 is also responsible for the phosphorylation and activation of DBL (an exchange factor for Rho) and Ras/GRF1 (an exchange factor for Ras).
Unit Definition
One unit is defined as the amount of enzyme that will phosphorylate 1 pmol of Tyr substrate per minute at pH 7.4 and 30°C.
Physical form
Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Involvement of Activated Cdc42 Kinase1 in Colitis and Colorectal Neoplasms.
Lv C, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 4794-4794 (2016)
The tyrosine kinase ACK1 associates with clathrin-coated vesicles through a binding motif shared by arrestin and other adaptors.
Teo M, et al.
The Journal of Biological Chemistry, 276, 18392-18392 (2001)
Ack1 mediates Cdc42-dependent cell migration and signaling to p130Cas.
Modzelewska K, et al.
The Journal of Biological Chemistry, 281, 37527-37527 (2006)
Ack1 overexpression promotes metastasis and indicates poor prognosis of hepatocellular carcinoma.
Lei X, et al.
Oncotarget, 6, 40622-40622 (2015)
Phosphorylation of WASP by the Cdc42-associated kinase ACK1: dual hydroxyamino acid specificity in a tyrosine kinase.
Yokoyama N, et al.
The Journal of Biological Chemistry, 280, 42219-42219 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service