SML3668
Ceralifimod hydrochloride
≥95% (HPLC)
Synonym(s):
1-((6-(2-Methoxy-4-propylbenzyloxy)-1-methyl-3,4-dihydronaphthalen-2-yl)methyl)azetidine-3-carboxylic acid hydrochloride, 1-[[3,4-Dihydro-6-[(2-methoxy-4-propylphenyl)methoxy]-1-methyl-2-naphthalenyl]methyl]-3-azetidinecarboxylic acid hydrochloride, 1-[[6-[(2-Methoxy-4-propylbenzyl)oxy]-1-methyl-3,4-dihydro-2-naphthalenyl]methyl]azetidine-3-carboxylic acid hydrochloride, ONO-4641 hydrochloride
About This Item
Quality Level
Assay
≥95% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
N4(CC(C4)C(=O)O)CC1=C(c2c(cc(cc2)OCc3c(cc(cc3)CCC)OC)CC1)C
InChI
1S/C27H33NO4/c1-4-5-19-6-7-22(26(12-19)31-3)17-32-24-10-11-25-18(2)21(9-8-20(25)13-24)14-28-15-23(16-28)27(29)30/h6-7,10-13,23H,4-5,8-9,14-17H2,1-3H3,(H,29,30)
InChI key
QDDQIPUKAXBMBX-UHFFFAOYSA-N
Biochem/physiol Actions
Caution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service