Skip to Content
Merck
All Photos(1)

Documents

SML3066

Sigma-Aldrich

PFI-6N

≥98% (HPLC)

Synonym(s):

N-Cyclopentyl-5-(4-(dimethylcarbamoyl)-3-hydroxyphenyl)-N-methylisoxazole-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O4
CAS Number:
Molecular Weight:
357.40
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=C(C=C(C=C1)C2=CC(C(N(C3CCCC3)C)=O)=NO2)O)N(C)C

Biochem/physiol Actions

PFI-6N is a structurally similar negative control compound for PFI-6 a potent and selective inhibitor of MLLT1/3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service