Skip to Content
Merck
All Photos(1)

Key Documents

SML2961

Sigma-Aldrich

GRL-0617

≥98% (HPLC)

Synonym(s):

5-Amino-2-methyl-N-[(1R)-1-(1-naphthalenyl)ethyl]benzamide, GRL 0617, GRL0617

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O
CAS Number:
Molecular Weight:
304.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -72.0 to -82.0°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(CCc2c3c(ccc2)cccc3)C(=O)c1c(ccc(c1)N)C

InChI

1S/C20H20N2O/c1-14-9-10-17(21)13-19(14)20(23)22-12-11-16-7-4-6-15-5-2-3-8-18(15)16/h2-10,13H,11-12,21H2,1H3,(H,22,23)

InChI key

SVHZQKLXYGYSMU-UHFFFAOYSA-N

Biochem/physiol Actions

GRL-0617 is a cell penetrant non-cytotoxic and potent inhibitor of the SARS-CoV -2 papain-like protease (SCoV2-PLpro) and SCoV-PLpro. GRL-0617 reversibly binds to SCoV2-PLpro and disrupts its interaction with Interferon-stimulated gene 15 (ISG15). GRL-0617 poorly affects (> 100 μM) the activities of MERS PLpro, a series of cysteine proteases, including the human deubiquitinating enzymes HAUSP, USP18, UCH-L1, UCH-L3, and a papain-like protease (PLP2) from the human coronavirus NL63. GRL-0617 suppress SARS-CoV-2 infection and promote anti-viral immunity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiira Ratia et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(42), 16119-16124 (2008-10-15)
We report the discovery and optimization of a potent inhibitor against the papain-like protease (PLpro) from the coronavirus that causes severe acute respiratory syndrome (SARS-CoV). This unique protease is not only responsible for processing the viral polyprotein into its functional
Donghyuk Shin et al.
Nature, 587(7835), 657-662 (2020-07-30)
The papain-like protease PLpro is an essential coronavirus enzyme that is required for processing viral polyproteins to generate a functional replicase complex and enable viral spread1,2. PLpro is also implicated in cleaving proteinaceous post-translational modifications on host proteins as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service