Skip to Content
Merck
All Photos(1)

Key Documents

SML2754

Sigma-Aldrich

Z3670677764

≥98% (HPLC)

Synonym(s):

N-[[5-Methoxy-2-(1-methylethyl)phenyl]methyl]-3-pyridinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€51.30
25 MG
€209.00

€51.30


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€51.30
25 MG
€209.00

About This Item

Empirical Formula (Hill Notation):
C16H20N2O
CAS Number:
Molecular Weight:
256.34
UNSPSC Code:
12352200
NACRES:
NA.77

€51.30


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(Cc2c(ccc(c2)OC)C(C)C)c1cnccc1

InChI

1S/C16H20N2O/c1-12(2)16-7-6-15(19-3)9-13(16)10-18-14-5-4-8-17-11-14/h4-9,11-12,18H,10H2,1-3H3

InChI key

ZGOCFOSBMLFDAA-UHFFFAOYSA-N

Biochem/physiol Actions

Z3670677764 is an inactive control for selective human melatonin MT2 receptor agonist UCSF4226 (ZINC128734226).
inactive control for selective human melatonin MT2 receptor agonist UCSF4226

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reed M Stein et al.
Nature, 579(7800), 609-614 (2020-02-11)
The neuromodulator melatonin synchronizes circadian rhythms and related physiological functions through the actions of two G-protein-coupled receptors: MT1 and MT2. Circadian release of melatonin at night from the pineal gland activates melatonin receptors in the suprachiasmatic nucleus of the hypothalamus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service