Skip to Content
Merck
All Photos(1)

Key Documents

SML2523

Sigma-Aldrich

Pramlintide acetate salt

≥95% (HPLC)

Synonym(s):

25,28,29 TriPro-amylin (human) acetate, AC 0137, AC 137, AC0137, AC137, TriPro-amylin acetate, [Pro25,28,29]-amylin (human) acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€169.00
25 MG
€683.00

€169.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€169.00
25 MG
€683.00

About This Item

Empirical Formula (Hill Notation):
C171H267N51O53S2
CAS Number:
Molecular Weight:
3949.39
UNSPSC Code:
12352200
NACRES:
NA.77

€169.00


Available to ship on17 April 2025Details


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

SC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)CS)CC(=O)N)[C@H](O)C)C)[C@H](O)C)C(=O)N[C@@H](C)C(=O)N[C@@H]([C@H](O)C)C(=O)N[C@@H](CCC(=O)N)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@

InChI

1S/C171H269N51O53S2/c1-21-81(12)130(163(268)207-110(56-78(6)7)169(274)222-53-33-42-118(222)170(275)221-52-32-41-117(221)160(265)219-135(89(20)230)167(272)206-109(66-125(180)238)151(256)212-128(79(8)9)161(266)186-68-126(239)192-111(70-223)154(259)203-107(64-123(178)236)152(257)218-134(88(19)229)166(271)195-98(136(181)241)57-92-43-45-94(231)46-44-92)214-159(264)116-40-31-51-220(116)127(240)69-187-141(246)101(58-90-34-24-22-25-35-90)199-148(253)105(62-121(176)234)201-149(254)106(63-122(177)235)202-155(260)112(71-224)209-156(261)113(72-225)208-146(251)103(60-93-67-184-75-188-93)205-162(267)129(80(10)11)213-150(255)100(55-77(4)5)198-145(250)102(59-91-36-26-23-27-37-91)200-147(252)104(61-120(175)233)196-137(242)82(13)189-144(249)99(54-76(2)3)197-142(247)96(39-30-50-185-171(182)183)193-143(248)97(47-48-119(174)232)194-165(270)132(86(17)227)215-138(243)83(14)190-157(262)114(73-276)211-168(273)133(87(18)228)216-139(244)84(15)191-164(269)131(85(16)226)217-153(258)108(65-124(179)237)204-158(263)115(74-277)210-140(245)95(173)38-28-29-49-172/h22-27,34-37,43-46,67,75-89,95-118,128-135,223-231,276-277H,21,28-33,38-42,47-66,68-74,172-173H2,1-20H3,(H2,174,232)(H2,175,233)(H2,176,234)(H2,177,235)(H2,178,236)(H2,179,237)(H2,180,238)(H2,181,241)(H,184,188)(H,186,266)(H,187,246)(H,189,249)(H,190,262)(H,191,269)(H,192,239)(H,193,248)(H,194,270)(H,195,271)(H,196,242)(H,197,247)(H,198,250)(H,199,253)(H,200,252)(H,201,254)(H,202,260)(H,203,259)(H,204,263)(H,205,267)(H,206,272)(H,207,268)(H,208,251)(H,209,261)(H,210,245)(H,211,273)(H,212,256)(H,213,255)(H,214,264)(H,215,243)(H,216,244)(H,217,258)(H,218,257)(H,219,265)(H4,182,183,185)/t81-,82-,83-,84-,85+,86+,87+,88+,89+,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,128-,129-,130-,131-,132-,133-,134-,135-/m0/s1

InChI key

NRKVKVQDUCJPIZ-MKAGXXMWSA-N

Biochem/physiol Actions

Pramlintide (AC0137, AC137, triPro-amylin) is a synthetic antidiabetic peptide that corresponds to human amylin (aa34-70) with proline substitutions at N25, S28 and S29 to limit the self-aggregation seen with native amylin. Pramlintide augments endogenous amylin-mediated glycemic control (blood glucose regulation) in vivo by delaying gastric emptying and inhibiting the secretion of glucagon, a catabolic hormone that opposes the effects of insulin and amylin.
Synthetic amylin (aa34-70) analog with in vivo antidiabetic efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service