Skip to Content
Merck
All Photos(1)

Key Documents

SML2448

Sigma-Aldrich

PK4C9

≥98% (HPLC)

Synonym(s):

Homocarbonyltopsentin, [2-[(6-Hydroxy-1H-indol-3-yl)carbonyl]-1H-imidazol-4-yl]-1H-indol-3-yl--methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€117.00
25 MG
€375.00

€117.00


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€117.00
25 MG
€375.00

About This Item

Empirical Formula (Hill Notation):
C21H14N4O3
CAS Number:
Molecular Weight:
370.36
UNSPSC Code:
12352200
NACRES:
NA.77

€117.00


Available to ship on11 April 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c(nc(c1)C(=O)c4c5c([nH]c4)cccc5)C(=O)c2c3c([nH]c2)cc(cc3)O

InChI

1S/C21H14N4O3/c26-11-5-6-13-15(9-23-17(13)7-11)20(28)21-24-10-18(25-21)19(27)14-8-22-16-4-2-1-3-12(14)16/h1-10,22-23,26H,(H,24,25)

InChI key

DRCVQVIMGSWRLN-UHFFFAOYSA-N

Biochem/physiol Actions

PK4C9 is a splice modulator of survival of motor neuron gene SMN2 that increases production of full-length SMN protein. Spinal muscular atrophy (SMA) is a motor neuron disease caused by deficiency in SMN protein resulting from loss of expression of the SMN1 gene. The related SMN2 gene can compensate, but polymorphism in SMN2 often results in altered splicing and exclusion of exon 7, which is required for a full-length SMN transcript. PK4C9 binds to pentaloop conformations of the stem-loop RNA structure TSL2, a cis-regulatory element for E7 inclusion, and promotes a shift to triloop conformations that display enhanced E7 splicing. In SMA cells, PK4C9 increased E7 inclusion by 40% accompanied by a 1.5-fold increase in SMN protein, a level shown to reverse SMA phenotypes in mice models.
Splice modulator of survival of motor neuron gene SMN2 that increases production of full-length SMN protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amparo Garcia-Lopez et al.
Nature communications, 9(1), 2032-2032 (2018-05-26)
Modification of SMN2 exon 7 (E7) splicing is a validated therapeutic strategy against spinal muscular atrophy (SMA). However, a target-based approach to identify small-molecule E7 splicing modifiers has not been attempted, which could reveal novel therapies with improved mechanistic insight.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service