Skip to Content
Merck
All Photos(1)

Documents

SML2393

Sigma-Aldrich

XL019

≥98% (HPLC)

Synonym(s):

(2S)-N-[4-[2-[[4-(4-Morpholinyl)phenyl]amino]-4-pyrimidinyl]phenyl]-2-pyrrolidinecarboxamide, (S)-N-(4-(2-(4-Morpholinophenylamino)pyrimidin-4-yl)phenyl)pyrrolidine-2-carboxamide, N-(4-{2-[(4-Morpholin-4-ylphenyl)amino]pyrimidin-4-yl}phenyl)-L-prolinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N6O2
CAS Number:
Molecular Weight:
444.53
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(C=C1)C2=NC(NC3=CC=C(C=C3)N4CCOCC4)=NC=C2)[C@H]5NCCC5

InChI

1S/C25H28N6O2/c32-24(23-2-1-12-26-23)28-19-5-3-18(4-6-19)22-11-13-27-25(30-22)29-20-7-9-21(10-8-20)31-14-16-33-17-15-31/h3-11,13,23,26H,1-2,12,14-17H2,(H,28,32)(H,27,29,30)/t23-/m0/s1

InChI key

ISOCDPQFIXDIMS-QHCPKHFHSA-N

Related Categories

Biochem/physiol Actions

XL019 is an ATP-binding site-targeting, orally active JAK2 subtype-selective Janus kinase inhibitor (IC50 in nM = 2.2/JAK2 vs. 134.3/JAK1, 214.2/JAK3, 348.3/TYK2) with much reduced or little activity against 116 other kinases (IC50 in nM = 125.4/PDGFRB, 139.7/FLT3, 225.8/c-KIT, 313.8/p70S6K, 370/MLK1, 375.4/IKKbeta, 483.6/KDR, 546.7/PDGFRA, 554.5/FLT4, 910.5/FLT; IC50 >1 μM toward remaining 106 kinases), CYP (1A2, 2C9, 2D6, 3A4 IC50 ≥20 μM), hERG (IC50 = 16 μM), and P-glycoprotein (IC50 >20 μM). XL019 effectively inhibits cellular STAT1/STAT3 phosphorylation both in HEL92.1.7 cultures (IC50 in nM = 386.4/pStat1 and 695/pStat3) and in HEL92.1.7 xenograft-derived tumor in mice in vivo (oral ED50 in mg/kg = 42/pStat1 and 210/pStat3) with significant HEL92.1.7 tumor suppression efficacy in mice (by 60% and 70% on day 14, respectively, with 200 and 300 mg/kg bid p.o. dosage).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy Forsyth et al.
Bioorganic & medicinal chemistry letters, 22(24), 7653-7658 (2012-11-07)
We report the discovery of a series of 4-aryl-2-aminoalkylpyrimidine derivatives as potent and selective JAK2 inhibitors. High throughput screening of our in-house compound library led to the identification of hit 1, from which optimization resulted in the discovery of highly
Charles G Mullighan et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(23), 9414-9418 (2009-05-28)
Pediatric acute lymphoblastic leukemia (ALL) is a heterogeneous disease consisting of distinct clinical and biological subtypes that are characterized by specific chromosomal abnormalities or gene mutations. Mutation of genes encoding tyrosine kinases is uncommon in ALL, with the exception of
Marilyn M Giacomini et al.
Drug metabolism and disposition: the biological fate of chemicals, 45(1), 76-85 (2016-11-03)
Inhibition of thiamine transporters has been proposed as a putative mechanism for the observation of Wernicke's encephalopathy and subsequent termination of clinical development of fedratinib, a Janus kinase inhibitor (JAKi). This study aimed to determine the potential for other JAKi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service