Skip to Content
Merck
All Photos(1)

Key Documents

SML2344

Sigma-Aldrich

SJB3-019A

≥98% (HPLC)

Synonym(s):

2-(Pyridin-3-yl)napthalo(-2,3-d)oxazole-4,9-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€50.12
25 MG
€231.00

€50.12

List Price€71.60Save 30%

Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€50.12
25 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C16H8N2O3
CAS Number:
Molecular Weight:
276.25
UNSPSC Code:
12352200
NACRES:
NA.77

€50.12

List Price€71.60Save 30%

Available to ship on15 April 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

potent USP1 inhibitor that induces ID1 degradation, upregulation of p21 and cytoxicity in K562 and other leukemic cell lines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Mistry et al.
Molecular cancer therapeutics, 12(12), 2651-2662 (2013-10-17)
Inhibitor of DNA binding 1 (ID1) transcription factor is essential for the proliferation and progression of many cancer types, including leukemia. However, the ID1 protein has not yet been therapeutically targeted in leukemia. ID1 is normally polyubiquitinated and degraded by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service