SML2123
BRF110
≥98% (HPLC)
Synonym(s):
4-[Methyl[2-phenyl-5-(2-propen-1-yl)-6-(trifluoromethyl)-4-pyrimidinyl]amino]benzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H18F3N3O2
CAS Number:
Molecular Weight:
413.39
UNSPSC Code:
12352200
NACRES:
NA.77
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=CC=C(N(C2=NC(C3=CC=CC=C3)=NC(C(F)(F)F)=C2CC=C)C)C=C1)O
Biochem/physiol Actions
BRF110 is a brain penetrant, potent and selective agonist of nuclear receptor-related 1 (Nurr1):Retinoid X receptor α (RXRα) heterodimer. BRF110 is a neuroprotective agent that prevents dopaminergic neuron demise and striatal dopaminergic denervation in vivo against PD-causing toxins. It induced dopamine biosynthesis in vivo. BRF110 chronic dosing does not induce dyskinesia in mice model.
brain penetrant, potent and selective agonist of Nurr1:RXRα heterodimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service